Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZGPAT, Chicken, Polyclonal Antibody, Abnova™

Catalog No. 89127376 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-127-376 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-127-376 Supplier Abnova Corporation Supplier No. PAB25752
Add to Cart
Add to Cart

Chicken polyclonal antibody raised against synthetic peptide of ZGPAT.

Specifications

Antigen ZGPAT
Applications ELISA, Immunofluorescence, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Description Chicken polyclonal antibody raised against synthetic peptide of ZGPAT.
Dilution Western Blot (1 ug/mL) Immunofluorescence (20 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.02% sodium azide)
Gene ZGPAT
Gene Alias GPATC6/GPATCH6/KIAA1847/MGC44880/ZC3H9/ZC3HDC9
Gene Symbols ZGPAT
Host Species Chicken
Immunogen A synthetic peptide corresponding to 15 amino acids at C-terminus of human ZGPAT.
Purification Method Peptide affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84619
Test Specificity Multiple isoforms of ZGPAT are known to exist.
Target Species Human
Content And Storage Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgY
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.