Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chicken Caronte Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF1051
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF1051 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF1051 Supplier R&D Systems Supplier No. BAF1051
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Caronte Polyclonal specifically detects Caronte in Chicken samples. It is validated for Western Blot.

Specifications

Antigen Caronte
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL
Gene Accession No. NP_990154
Gene Alias Chicken Caronte
Gene Symbols CER1
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant chicken Caronte Asp16-Asn272 Accession # NP_990154
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9350
Test Specificity Detects chicken Caronte in Western blots. In this format, less than 1% cross-reactivity with recombinant human DAN and recombinant mouse Gremlin is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Chicken
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.