Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chicken IgG Isotype Control, FITC, Novus Biologicals™
SDP

Catalog No. NBP196860 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP196860 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP196860 Supplier Novus Biologicals Supplier No. NBP196860
Add to Cart
Add to Cart

Chicken Antibody

IgG specifically detects IgG in Chicken samples. It is validated for SDS-Page.

Specifications

Antigen IgG
Applications SDS-Page
Conjugate Fluorescein
Dilution SDS-Page 1:100 - 1:2000
Formulation Lyophilized from 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free with 0.01% Sodium Azide
Gene Alias Immunoglobulin G, ImmunoglobulinG
Gene Symbols IGHG1
Host Species Chicken
Purification Method Multi-step
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Isotype Control (Primary)
Gene ID (Entrez) 3500
Test Specificity Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Fluorescein, anti-Chicken IgG and anti-Chicken Serum.
Reconstitution Reconstitute with 1.0 ml deionized water (or equivalent).
Target Species Chicken
Content And Storage Store lyophilized antibody at 4°C in the dark. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles.
Form Lyophilized
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.