Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chicken Netrin-2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB127
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB127 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB127 Supplier R&D Systems Supplier No. MAB127
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Netrin-2 Monoclonal specifically detects Netrin-2 in Chicken samples. It is validated for Western Blot.

Specifications

Antigen Netrin-2
Applications Western Blot
Classification Monoclonal
Clone 74506
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q90923
Gene Alias Netrin2, NTN2L
Gene Symbols NTN3
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant chicken Netrin-2 Ala16-Pro581 Accession # Q90923
Purification Method Protein A or G purified from ascites
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4917
Test Specificity Detects chicken Netrin-2 in direct ELISAs and Western blots.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Chicken
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.