Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chikungunya E1 Mouse, Clone: 1D5, Abnova™

Catalog No. 89933970 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-933-970 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-933-970 Supplier Abnova Corporation Supplier No. MAB12423

Mouse monoclonal antibody raised against Chikungunya E1.

Specifications

Antigen Chikungunya E1
Applications ELISA
Classification Monoclonal
Clone 1D5
Conjugate Unconjugated
Formulation Lyophilized from PBS, pH 7.2 (0.09% sodium azide)
Gene Symbols CHIKVgp2
Host Species Mouse
Immunogen Native purified Chikungunya E1.
Purification Method Antigen Affinity Purification
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 956308
Target Species Virus
Content And Storage Store at 4°C on dry atmosphere. After reconstitution with 1 mL of deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.