Learn More
Chikungunya E1 Protein Monoclonal Antibody (CHK-166), Functional Grade, In Vivo PLATINUM™, Leinco Technologies™

Mouse Monoclonal Antibody
Supplier: Leinco Technologies Inc C479100MG


Description
Specificity: CHK-166 activity is directed against CHIKV E1. Endotoxin Level: <0.5 EU/mg as determined by the LAL method. Purity: ≥98% monomer by analytical SEC and >95% by SDS Page. Preparation: Functional grade preclinical antibodies are manufactured in an animal free facility using in vitro cell culture techniques and are purified by a multi-step process including the use of protein A or G to assure extremely low levels of endotoxins, leachable protein A or aggregates. Pathogen Testing: To protect mouse colonies from infection by pathogens and to assure that experimental preclinical data is not affected by such pathogens, all of Leinco Purified Functional PLATINUM™ antibodies are tested and guaranteed to be negative for all pathogens in the IDEXX IMPACT I Mouse Profile. Storage: Functional grade preclinical antibodies may be stored sterile as received at 2-8C for up to one month. For longer term storage, aseptically aliquot in working volumes without diluting and store at -80C. Avoid Repeated Freeze Thaw Cycles.
Chikungunya E1 Protein Monoclonal antibody specifically detects Chikungunya E1 Protein in Virus samples. It is validated for ELISA, Flow Cytometry, Functional Assay, NeutralizationSpecifications
Chikungunya E1 Protein | |
Monoclonal | |
≥5.0 mg/mL | |
PBS with no preservative; pH 7.2-7.4 | |
Mouse | |
Protein A/G | |
RUO | |
Virus | |
Antibody | |
IgG2c |
ELISA, Flow Cytometry, Functional Assay, Neutralization | |
CHK-166 | |
Functional Grade | |
Chikungunya Virus | |
Chikungunya E1 protein | |
100 mg | |
Primary | |
4°C short term, -80°C long term | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.