Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chikungunya E2 Protein Monoclonal Antibody (CHK152), Functional Grade, In Vivo GOLD™, Leinco Technologies™
SDP

Catalog No. 502544227
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-4227 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-4227 Supplier Leinco Technologies Inc Supplier No. C450100MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specificity: Clone CHK152 binds to and shows mechanisms of neutralizing of the Chikungunya E2 protein. Endotoxin Level: <1.0 EU/μg as determined by the LAL method. Purity: ≥95% monomer by analytical SEC and >95% by SDS Page. Preparation: Functional grade preclinical antibodies are manufactured in an animal free facility using in vitro cell culture techniques and are purified by a multi-step process including the use of protein A or G to assure extremely low levels of endotoxins, leachable protein A or aggregates. Storage: Functional grade preclinical antibodies may be stored sterile as received at 2-8C for up to one month. For longer term storage, aseptically aliquot in working volumes without diluting and store at -80C. Avoid Repeated Freeze Thaw Cycles.

Chikungunya E2 Protein Monoclonal antibody specifically detects Chikungunya E2 Protein in Virus samples. It is validated for ELISA, Functional Assay, Immunohistochemistry, Neutralization

Specifications

Antigen Chikungunya E2 Protein
Applications ELISA, Functional Assay, Immunohistochemistry, Neutralization
Classification Monoclonal
Clone CHK152
Concentration ≥5.0 mg/mL
Conjugate Functional Grade
Formulation PBS with no preservative; pH 7.2-7.4
Host Species Mouse
Immunogen Chikungunya E2 protein
Purification Method Protein A/G
Quantity 100 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage 4°C short term, -80°C long term
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.