Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chikungunya Virus Antibody (3E7b), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 1 publication
$182.78 - $519.67
Specifications
| Antigen | Chikungunya virus |
|---|---|
| Clone | 3E7b |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Functional |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Functional Assay |
Description
Chikungunya Virus Monoclonal specifically detects Chikungunya Virus in Virus samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
| Chikungunya virus | |
| 1.0 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Functional Assay | |
| Unconjugated | |
| Mouse | |
| Immunology, Infections (Virus Bacteria and Parasites), Virology Bacteria and Parasites | |
| PBS with 0.02% Sodium Azide | |
| Live chikungunya virus | |
| Primary | |
| Both Anti-CHIKV [3E7b] and Anti-CHIKV [8A2c] bind strongly to native CHIKV surface and potently neutralize CHIKV replication. |
| 3E7b | |
| Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Functional | |
| Monoclonal | |
| Purified | |
| RUO | |
| Virus | |
| Chikungunya | |
| IgM | |
| Thiophilic Chromatography |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title