Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chikungunya Virus Antibody (6A11), Alexa Fluor™ 750, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP253111AF750

Catalog No. NP23111A70


Only null left
Add to Cart

Description

Description

Chikungunya Virus Monoclonal specifically detects Chikungunya Virus in Virus samples. It is validated for Western Blot, Immunofluorescence.
Specifications

Specifications

Chikungunya Virus
Monoclonal
Alexa Fluor 750
50 mM sodium borate with 0.05% sodium azide
Mouse
Protein A purified
Immunology, Infections (Virus Bacteria and Parasites), Virology Bacteria and Parasites
Virus
IgG2b
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
6A11
Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence
Chikungunya
Live Chikungunya Virus
0.1 mL
Primary
Store at 4°C in the dark.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.