Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chikungunya Virus Antibody (8A2c), Novus Biologicals™

Mouse Monoclonal Antibody
$196.10 - $499.50
Specifications
Antigen | Chikungunya virus |
---|---|
Clone | 8A2c |
Concentration | 1.0 mg/mL |
Dilution | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:10-1:500, Functional |
Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Functional Assay |
Description
Chikungunya Virus Monoclonal specifically detects Chikungunya Virus in Virus samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
Chikungunya virus | |
1.0 mg/mL | |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Functional Assay | |
Unconjugated | |
Mouse | |
Immunology, Infections (Virus Bacteria and Parasites), Virology Bacteria and Parasites | |
PBS with 0.02% Sodium Azide | |
Live chikungunya virus | |
Primary |
8A2c | |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:10-1:500, Functional | |
Monoclonal | |
Purified | |
RUO | |
Virus | |
Chikungunya | |
IgM | |
Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title