Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chikungunya Virus Antibody (E26C4), Novus Biologicals™

Human Monoclonal Antibody
Supplier: Novus Biologicals DDX9102P100
Description
Chikungunya Virus Monoclonal specifically detects Chikungunya Virus in Virus samples. It is validated for Chromatin Immunoprecipitation, Immunocytochemistry/Immunofluorescence, Neutralization, Block/Neutralize.Specifications
Chikungunya virus | |
Monoclonal | |
0.5 mg/mL | |
Immunocytochemistry/Immunofluorescence 1:10-1:2000, Neutralization, Chromatin Immunoprecipitation (ChIP), Block/Neutralize | |
Human | |
Ion exchange chromatography | |
RUO | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Immunocytochemistry, Immunofluorescence, Neutralization, ChIP Assay, Blocking Assay | |
E26C4 | |
Unconjugated | |
Chikungunya | |
Chikungunya virus was used as immunogen. | |
0.1 mg | |
Immunology, Infections (Virus Bacteria and Parasites), Virology Bacteria and Parasites | |
Human, Monkey, Virus | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction