Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Chikungunya Virus Monoclonal Antibody (D51Q)
GREENER_CHOICE

Catalog No. PIMA518180
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA518180 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA518180 Supplier Invitrogen™ Supplier No. MA518180
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody is specific to Chikungunya virus and does not cross-react with other Alphaviruses Eastern Equine Encephalitis, Western Equine Encephalitis, Venezuelan Equine Encephalitis or other Arboviruses. Purity is >90%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Chikungunya Virus
Applications ELISA, Immunocytochemistry
Classification Monoclonal
Clone D51Q
Concentration 100 μg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.2
Gene Alias CHIKV
Host Species Mouse
Immunogen Chikungunya virus native protein.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.