Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chikungunya virus Mouse anti-Virus, Clone: 3E7b, MilliporeSigma™

Catalog No. MABF205225 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF2052-25 25 μL
MABF2052-100 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABF2052-25 Supplier MilliporeSigma Supplier No. MABF205225UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Chikungunya virus, clone 3E7b is a mouse monoclonal antibody that detects E2 protein of Chikungunya virus and has been tested for use in ELISA, Immunocytochemistry, Neutralizing asssay, and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Chikungunya virus
Applications ELISA, Immunocytochemistry, Neutralization, Western Blot
Classification Monoclonal
Clone 3E7b
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Host Species Mouse
Immunogen Chikungunya virus
Purification Method Purified by ion-exchange chromatography
Quantity 25 μL
Regulatory Status RUO
Research Discipline Inflammation and Immunology
Primary or Secondary Primary
Test Specificity Clone 3E7b detects Chikungunya virus in cells infected with this virus.
Target Species Virus
Content And Storage Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Form Purified
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.