Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chikungunya Virus nsp3 Antibody (HL2479) - Azide and BSA Free, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP325411
Description
Chikungunya Virus nsp3 Monoclonal antibody specifically detects Chikungunya Virus nsp3 in Virus samples. It is validated for Western BlotSpecifications
Chikungunya Virus nsp3 | |
Monoclonal | |
Unconjugated | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Virus | |
Purified |
Western Blot | |
HL2479 | |
Western Blot 1:5000-1:50000 | |
CHIKV nsp3, Non-structural protein 3 | |
Recombinant fragment of Chikungunya Virus nsp3. | |
100 μL | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction