Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Chikungunya Virus NSP3 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA5143451

Catalog No. FEPA5143451


Only null left
Add to Cart

Description

Description

Store as concentrated solution. Centrifuge briefly prior to opening vial.

Chikungunya virus (CHIKV) is a re-emerging Alphavirus causing fever, joint pain, skin rash, arthralgia, and occasionally death The non-structural protein 3 (nsP3) plays an essential role in RNA replication The nsP3 alphavirus unique domain (AUD) is a multi-functional domain It is not only a critical determinant of both cell and species-specificity, but also plays roles in virus genome replication and assembly [PMID: 30668592].
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Chikungunya Virus NSP3
Polyclonal
Unconjugated
CHIKV NSP3; NsP3 protein
Recombinant protein encompassing a sequence within the center region of Chikungunya virus nsP3. The exact sequence is proprietary.
100 μL
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
1.19 mg/mL
PBS with 20% glycerol and 0.025% Proclin 300; pH 7
Rabbit
Protein A
RUO
Virus
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.