Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chikungunya Virus VLP Mouse anti-Virus, Clone: AG7-2-A7-E10, Novus Biologicals™

Mouse Monoclonal Antibody
$556.26 - $1381.04
Specifications
| Antigen | Chikungunya Virus VLP |
|---|---|
| Clone | AG7-2-A7-E10 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot, ELISA |
| Applications | Western Blot, ELISA |
Description
Chikungunya Virus VLP Monoclonal antibody specifically detects Chikungunya Virus VLP in Virus samples. It is validated for Western Blot, ELISASpecifications
| Chikungunya Virus VLP | |
| 1 mg/mL | |
| Western Blot, ELISA | |
| Unconjugated | |
| Mouse | |
| Virus | |
| An equal mix of Chikungunya, Mayaro and O'nyong'nyong VLPs (comprising E1, E2 and capsid proteins), produced in HEK293 cells | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| AG7-2-A7-E10 | |
| Western Blot, ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS pH 7.4 | |
| IgG | |
| Protein G purified | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title