Learn More
Invitrogen™ CHIL3 Monoclonal Antibody (281926)

Description
Reconstitute at 0.5 mg/mL in sterile PBS.
Specifications
Specifications
| Antigen | CHIL3 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 281926 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5% trehalose and No Preservative |
| Gene | Chil3 |
| Gene Accession No. | O35744 |
| Gene Alias | AI505981; beta-N-acetylhexosaminidase Ym1; Chi3l3; Chil3; chitinase 3-like 3; chitinase-3-like protein 3; chitinase-like 3; chitinase-like protein 3; ECF-L; eosinophil chemotactic cytokine; eosinophil chemotactic factor-L; secreted protein Ym1; Ym1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.