Learn More
Ergodyne™ Chill-Its™ 6700FR Evaporative FR Cooling Tie Bandana
FR Evaporative Cooling Bandana featured activated polymers that provide cooling relief and FR protection.
$16.97
Specifications
Material | Modacrylic Cotton Blend |
---|---|
For Use With (Application) | Construction, Maintenance, Landscaping, Material Handling |
Catalog Number | Mfr. No. | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Price | Quantity & Availability | |||||
19-819-584
|
Ergodyne™
12601 |
Lime |
Each for $16.97
|
|
|||||
19-819-586
|
Ergodyne™
12607 |
Blue |
Each for $16.97
|
|
|||||
Description
- Modacrylic cotton blend fabric
- Fabric meets ASTM F1506 performance requirements (ATPV rating of 7.1 cal/cm2); classified CAT1 per NFPA 70E.
- Meets ASTM F2302 for flame resistance
- Soak in water 2-5 minutes to activate
- Remains hydrated for up to 4 hours
Specifications
Modacrylic Cotton Blend | |
Construction, Maintenance, Landscaping, Material Handling |