Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chinese Hamster PLBD2 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
$177.10
Specifications
Antigen | PLBD2 |
---|---|
Clone | 2751B |
Dilution | ELISA |
Applications | ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB106921SP
![]() |
R&D Systems
MAB106921SP |
25 μg |
Each for $177.10
|
|
|||||
MAB106921
|
R&D Systems
MAB106921 |
100 μg | N/A | N/A | N/A | ||||
Description
PLBD2 Monoclonal antibody specifically detects PLBD2 in Chinese Hamster samples. It is validated for ELISASpecifications
PLBD2 | |
ELISA | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
LAMA-like protein 2,76 kDa protein, lamina ancestor homolog 2, mannose-6-phosphate protein associated protein p76, P76, phospholipase B domain containing 2, phospholipase B domain-containing protein 2, phospholipase B-like 2 32 kDa form, phospholipase B-like 2 45 kDa form, PLB homolog 2, putative phospholipase B-like 2 | |
Recombinant chinese hamster PLBD-2, Met1-Asp585, Accession # XP_003510860 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
2751B | |
ELISA | |
Unconjugated | |
Rabbit | |
Chinese Hamster | |
XP_003510860 | |
196463 | |
IgG | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title