Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chinese Hamster PLBD2 Antibody, R&D Systems™

Catalog No. MAB10692
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB10692 100 μg
MAB10692SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB10692 Supplier R&D Systems Supplier No. MAB10692
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

PLBD2 Monoclonal antibody specifically detects PLBD2 in Chinese Hamster samples. It is validated for nan

Specifications

Antigen PLBD2
Classification Monoclonal
Clone 2751C
Conjugate Unconjugated
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS.
Gene Alias LAMA-like protein 2,76 kDa protein, lamina ancestor homolog 2, mannose-6-phosphate protein associated protein p76, P76, phospholipase B domain containing 2, phospholipase B domain-containing protein 2, phospholipase B-like 2 32 kDa form, phospholipase B-like 2 45 kDa form, PLB homolog 2, putative phospholipase B-like 2
Host Species Rabbit
Immunogen Recombinant Chinese Hamster PLBD-2, Met1-Asp585, Accession # XP_003510860
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 196463
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Chinese Hamster
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.