Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CHIP/STUB1 Antibody (034), DyLight 488, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290116G
Description
CHIP/STUB1 Monoclonal specifically detects CHIP/STUB1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
CHIP/STUB1 | |
Monoclonal | |
DyLight 488 | |
Antigen NY-CO-7, Carboxy terminus of Hsp70-interacting protein, CHIPSTIP1 homology and U box-containing protein 1, CLL-associated antigen KW-8, E3 ubiquitin-protein ligase CHIP, EC 6.3.2.-, heat shock protein A binding protein 2 (c-terminal), HSPABP2, NY-CO-7, SDCCAG7, serologically defined colon cancer antigen 7, STIP1 homology and U-box containing protein 1, STIP1 homology and U-box containing protein 1, E3 ubiquitin protein ligase, UBOX1 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human CHIP/STUB1 isoform 1 (Q9UNE7-1; Met 1-Tyr 303). | |
0.1 mL | |
Neuroscience | |
10273 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, Flow Cytometry, ELISA, Immunofluorescence | |
34 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction