Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CHIP/STUB1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10077316
Description
CHIP/STUB1 Polyclonal specifically detects CHIP/STUB1 in Human samples. It is validated for Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Western Blot (Negative).Specifications
CHIP/STUB1 | |
Polyclonal | |
Unconjugated | |
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
Antigen NY-CO-7, Carboxy terminus of Hsp70-interacting protein, CHIPSTIP1 homology and U box-containing protein 1, CLL-associated antigen KW-8, E3 ubiquitin-protein ligase CHIP, EC 6.3.2.-, heat shock protein A binding protein 2 (c-terminal), HSPABP2, NY-CO-7, SDCCAG7, serologically defined colon cancer antigen 7, STIP1 homology and U-box containing protein 1, STIP1 homology and U-box containing protein 1, E3 ubiquitin protein ligase, UBOX1 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Based on 100% sequence identity, this antibody is predicted to react with X. tropicalis, Panda, Orangutan, Rhesus Monkey, Gorilla, Chimpanzee, West Indian ocean coelacanth, Tasmanian devil and Northern white-cheeked gibbon. | |
Store at 4C. Do not freeze. |
Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot | |
1.0 mg/mL | |
Immunohistochemistry 1:10-1:500, Immunoprecipitation 2-5 ug/mg lysate, Immunohistochemistry-Paraffin 1:100-1:500, Western Blot (Negative) | |
NP_005852.2 | |
STUB1 | |
The immunogen recognized by this antibody maps to a region between residue 253 and 303 of human STIP1 homology and U-box containing protein 1 (carboxy terminus of Hsp70-interacting protein) using the numbering given in entry NP_005852.2 (GeneID 10273). | |
100 μL | |
Neuroscience | |
10273 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction