Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CHIP/STUB1 Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NB300982
Description
CHIP/STUB1 Polyclonal specifically detects CHIP/STUB1 in Human samples. It is validated for Western Blot, Peptide ELISA.Specifications
CHIP/STUB1 | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
Antigen NY-CO-7, Carboxy terminus of Hsp70-interacting protein, CHIPSTIP1 homology and U box-containing protein 1, CLL-associated antigen KW-8, E3 ubiquitin-protein ligase CHIP, EC 6.3.2.-, heat shock protein A binding protein 2 (c-terminal), HSPABP2, NY-CO-7, SDCCAG7, serologically defined colon cancer antigen 7, STIP1 homology and U-box containing protein 1, STIP1 homology and U-box containing protein 1, E3 ubiquitin protein ligase, UBOX1 | |
Goat | |
Affinity Purified | |
RUO | |
Primary | |
Cross reacts with Human. Expected from sequence similarity: Mouse, Rat, Dog and Cow. | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot, ELISA | |
0.5 mg/mL | |
Western Blot 0.3 - 1 ug/ml, Peptide ELISA Detection limit 1:64000 | |
NP_005852. | |
STUB1 | |
Peptide with sequence C-DAFISENGWVEDY corresponding to C-Terminus according to NP_005852.2. | |
0.1 mg | |
Neuroscience | |
10273 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction