Learn More
Description
For more information about how CHIR 99021 may be used, please see our protocols: Transdifferentiating Fibroblasts into Neurons, Culturing Transcriptomically Distinct Pluripotent mESCs, Converting Fibroblasts into Cardiomyocytes (9C Cocktail), Human iPSC-derived Myoblasts, Reprogramming MEFs into Skeletal Muscle Progenitors, Highly Efficient Generation of CiPSCs from MEFs, Generation of Kidney Organoids from hPSCs, Culturing Transcriptomically Distinct Pluripotent mESCs .
Research use only CHIR 99021 also available.
Specifications
Specifications
| Chemical Name or Material | 6-[[2-[[4-(2,4-Dichlorophenyl)-5-(5-methyl-1H-imidazol-2-yl)-2-pyrimidinyl]amino]ethyl]amino]-3-pyridinecarbonitrile |
| CAS | 252917-06-9 |
| Quantity | 10 mg |
| Product Line | Tocris |
| Target | GSK-3 Inhibitors |
| Molecular Formula | C22H18Cl2N8 |
| Purity | 99% |
| Regulatory Status | GMP |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
