Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ CHIR 99021
GSA_VAGREENER_CHOICESDP

Catalog No. 442350 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-235-0 50 mg
44-231-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 44-235-0 Supplier R&D Systems™ Supplier No. 4423/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Highly selective GSK-3 inhibitor

CHIR 99021 is a potent and highly selective inhibitor of glycogen synthase kinase 3 (GSK-3) (IC50 values are 6.7 and 10 nM for GSK-3β and GSK-3α respectively). Acts as Wnt activator. Exhibits >500-fold selectivity for GSK-3 over closely related kinases; also displays >800-fold selectivity against 45 additional enzymes and receptors. In combination with the irreversible inhibitor of LSD1 (Cat. No. 3852), CHIR 99021 enables reprogramming of mouse embryonic fibroblasts, transduced by Oct4 and Klf4 only, into iPSCs. Enhances mouse and human ESC self-renewal when used in combination with PD 0325901 (Cat. No. 4192). It promotes differentiation of hPSCs in cardiomyocytes and allows formation of extended pluripotent stem (EPS) cells in combination with (S)-(+)-Dimethindene maleate (Cat. No. 1425), tetracycline antibiotic (Cat. No. 3268) and human leukemia inhibitory factor. CHIR 99021 synthesized to cGMP guidelines also available. For more information on how to use CHIR 99021, see our protocols: Transdifferentiating Fibroblasts into Neurons, Culturing Transcriptomically Distinct Pluripotent mESCs, Converting Fibroblasts into Cardiomyocytes (9C Cocktail), Human iPSC-derived Myoblasts .

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Chemical Identifiers

CAS 252917-06-9
Molecular Formula C22H18Cl2N8

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 465.34
Purity >99%
Molecular Weight (g/mol) 465.34
Chemical Name or Material CHIR 99021
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.