Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ CHIRALPAK™ CBH HPLC Column, 5 μm Particle Size

Catalog No. 111005789 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Length (Metric):
10 cm
15 cm
Diameter (Metric):
2 mm
3 mm
4 mm
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Diameter (Metric)
11-100-5789 10 cm 2 mm
11-100-5818 10 cm 3 mm
11-100-5774 15 cm 3 mm
11-100-5819 10 cm 4 mm
11-100-5773 15 cm 4 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 11-100-5789 Supplier MilliporeSigma™ Supelco™ Supplier No. 58530AST
Add to Cart
Add to Cart
Request a Quote

CHIRALPAK™ CBH phase has cellobiohydrolase as the chiral selector immobilized on spherical 5 μm silica particles.

CHIRALPAK™ CBH phase has cellobiohydrolase as the chiral selector immobilized on spherical 5 μm silica particles. Used in reversed-phase mode, retention and enantioselectivity is regulated by changes of pH, buffer concentration and organic modifier. The mobile phases are mixtures of phosphate or acetate buffers and organic solvents such as 2-propanol or acetonitrile. The column is preferably used for the separation of enantiomers of basic drugs, particularly compounds containing one or more nitrogen atoms along with one or more hydrogen-bonding groups (alcohol, phenol, carbonyl, amide, ether, ester, etc.). Features: Direct reversed phase resolution of chiral molecules. Stable with a variety of organic modifiers. Available in analytical and semi-preparative sizes.
TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) 2 mm
Length (Metric) 10 cm
Particle Size 5 μm
Stationary Phase Cellobiohydrolase
Product Line CHIRALPAK
Matrix Cellobiohydrolase Phase
Phase Chiral
Technique Chiral HPLC

CHIRALPAK is a registered trademark of Daicel Corp.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.