Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chitin, Practical grade, MP Biomedicals™

Catalog No. p-4403646 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
10 g
1 kg

Chemical Identifiers

CAS 1398-61-4
Molecular Formula (C8H13NO5)nC16H28N2O11
Synonym chitin from shrimp shells, n-2r-2,4,5-trihydroxy-6-hydroxymethyl oxan-3-yl acetamide, chitin from shrimp shells, practical grade, powder, chitin from shrimp shells, vetec tm reagent grade, chitin from shrimp shells, practical grade, coarse flakes, chitin from shrimp shells, bioreagent, suitable for analysis of chitinase, purified powder

Specifications

Color White to Brown
Quantity 10 g
Formula Weight 770.8
Grade Practical
Physical Form Powder
Chemical Name or Material Chitin
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.