Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chitinase-1 (CHIT1) Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. ABF116825UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABF1168-25UL 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABF1168-25UL Supplier MilliporeSigma Supplier No. ABF116825UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Chitinase-1 (CHIT1) is a rabbit polyclonal that detects Chitinase-1 in human cells and has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Chitinase-1 (CHIT1)
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols CHIT1
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to 15 amino acids from the C-terminal region of human Chitinase-1.
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Inflammation and Immunology
Primary or Secondary Primary
Gene ID (Entrez) NP_003456.1.
Test Specificity This rabbit polyclonal antibody detects Chitinase-1 in human serum. It targets an epitope within 15 amino acids from the C-terminal region.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.