Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chitinase 3-like 1 Antibody (101), Alexa Fluor™ 750 , Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289852AF750
Description
Chitinase 3-like 1 Monoclonal antibody specifically detects Chitinase 3-like 1 in Human samples. It is validated for ELISASpecifications
Chitinase 3-like 1 | |
Monoclonal | |
Alexa Fluor 750 | |
AW208766, Brp39, Chi3l1, gp39 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human Chitinase 3-like 1 (NP_001267.2; Met1-Thr383). | |
0.1 mL | |
Cell Biology, Cytoskeleton Markers, Signal Transduction | |
1116 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
101 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction