Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chitinase 3-like 1 Mouse anti-Human, Clone: CL12241, Novus Biologicals™

Mouse Monoclonal Antibody
$366.50 - $607.50
Specifications
Antigen | Chitinase 3-like 1 |
---|---|
Clone | CL12241 |
Dilution | Western Blot 1 μg/mL, Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500 |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Description
Chitinase 3-like 1 Monoclonal antibody specifically detects Chitinase 3-like 1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
Chitinase 3-like 1 | |
Western Blot 1 μg/mL, Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
AW208766, Brp39, Chi3l1, gp39 | |
This antibody was developed against a synthetic peptide. Actual immunogen sequence is proprietary information. | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
CL12241 | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Cell Biology, Cytoskeleton Markers, Signal Transduction | |
PBS (pH 7.2), 40% glycerol | |
1116 | |
IgG2b | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title