Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chitinase 3-like 1 Rabbit anti-Human, Clone: 101, Novus Biologicals™

Rabbit Monoclonal Antibody
$353.50 - $560.50
Specifications
Antigen | Chitinase 3-like 1 |
---|---|
Clone | 101 |
Dilution | ELISA 1:5000-1:10000 |
Applications | ELISA |
Classification | Monoclonal |
Description
Chitinase 3-like 1 Monoclonal specifically detects Chitinase 3-like 1 in Human samples. It is validated for ELISA.Specifications
Chitinase 3-like 1 | |
ELISA 1:5000-1:10000 | |
Monoclonal | |
Purified | |
RUO | |
0.2 um filtered solution in PBS | |
1116 | |
IgG | |
Protein A purified |
101 | |
ELISA | |
Unconjugated | |
Rabbit | |
Human | |
AW208766, Brp39, Chi3l1, gp39 | |
Recombinant Human Chitinase 3-like 1 protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title