Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chitinase 3-like 1 Rabbit anti-Human, Mouse, Clone: 9P1P6, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | Chitinase 3-like 1 |
---|---|
Clone | 9P1P6 |
Applications | Western Blot, Immunofluorescence |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
Chitinase 3-like 1 Monoclonal antibody specifically detects Chitinase 3-like 1 in Human, Mouse samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
Chitinase 3-like 1 | |
Western Blot, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Cell Biology, Cytoskeleton Markers, Signal Transduction | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
1116 | |
IgG | |
Affinity purified |
9P1P6 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse | |
AW208766, Brp39, Chi3l1, gp39 | |
A synthetic peptide corresponding to a sequence within amino acids 284-383 of human Chitinase 3-like 1 (NP_001267.2). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title