Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chitinase 3-like 2 Antibody (213), PerCP, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290064PCP
Description
Chitinase 3-like 2 Monoclonal antibody specifically detects Chitinase 3-like 2 in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)Specifications
Chitinase 3-like 2 | |
Monoclonal | |
PerCP | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA, Immunohistochemistry (Paraffin) | |
213 | |
ELISA, Immunohistochemistry-Paraffin | |
chitinase 3-like 2, chitinase-3-like protein 2, Chondrocyte protein 39, YKL-39YKL39 | |
Rabbit immunized with purified, recombinant Human Chitinase 3-like 2 (Accession#: NP_003991.2, Met1-Leu390) | |
0.1 mL | |
Stem Cells | |
1117 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction