Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chitosan, 85% deacetylated

Catalog No. aaj6414318
Click to view available options
Quantity:
50 g
250 g
500 g

Chitosan, CAS # 9012-76-4, is a polymer derived from chitin. It is used in the laboratory research of medical material and biomedicine due to its demonstrated bactericidal, antibacterial, anticancer, and antiviral effects. | CAS: 9012-76-4 | C56H103N9O39 | 1526.464 g/mol

Chitosan is widely utilized in biomedical applications. It finds applications in plantations as natural seed treatment and a growth enhancer. It serves as a fining agent in winemaking and also prevents spoilage. It is involved in the field of medicine to reduce bleeding and as an antibacterial agent. It is used in self-healing coatings which are applied in industrial applications. It is also employed as a part of a filtration process to remove phosphorus, heavy minerals and oils from water.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Chitosan is a naturally derived polymer formed from the deacetylation of chitin
  • It is an alkaline polysaccharide with moisturizing and adsorption properties
  • Derivatives of chitosan are produced by chemically modifying chitosan-reactive functional groups

Application

  • Chitosan is used in the laboratory research of medical material and biomedicine due to its bactericidal, antibacterial, anticancer, and antiviral effects
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9012-76-4
Molecular Formula C56H103N9O39
Molecular Weight (g/mol) 1526.464
MDL Number MFCD00161512
InChI Key FLASNYPZGWUPSU-SICDJOISSA-N
Synonym chitosan, poliglusam, deacetylchitin, chicol, flonac c, flonac n, sea cure plus, kytex h, kytex m, kimitsu chitosan f
PubChem CID 71853
IUPAC Name methyl N-[(2S,3R,4R,5S,6R)-5-[(2S,3R,4R,5S,6R)-3-amino-5-[(2S,3R,4R,5S,6R)-3-amino-5-[(2S,3R,4R,5S,6R)-3-amino-5-[(2S,3R,4R,5S,6R)-3-amino-5-[(2S,3R,4R,5S,6R)-3-amino-5-[(2S,3R,4R,5S,6R)-3-amino-4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-4-hydroxy-6-(hy
SMILES COC(=O)NC1C(C(C(OC1OC2C(OC(C(C2O)N)OC3C(OC(C(C3O)N)O)CO)CO)CO)OC4C(C(C(C(O4)CO)OC5C(C(C(C(O5)CO)OC6C(C(C(C(O6)CO)OC7C(C(C(C(O7)CO)OC8C(C(C(C(O8)CO)OC9C(C(C(C(O9)CO)O)O)N)O)N)O)N)O)N)O)N)O)N)O

Specifications

Color White
Physical Form Powder
Solubility Information Soluble in acetic acid,adipic acid,formic acid,lactic acid,propionic acid and succinic acid.Insoluble in phosphoric acid,sulfuric acid,neutral and alkaline media.
Quantity 50 g
Chemical Name or Material Chitosan, 85% deacetylated
RTECSNumber FM6300000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.