Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chitosan, Powder, Spectrum™ Chemical
SDP

Catalog No. 18601571 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
5 kg
Packaging:
Poly Bottle
Poly Pail

C1569, 9012-76-4, (C6H11NO4)n

Spectrum™ Chemical Chitosan, Powder is a polysaccharide made by treating shrimp and other crustacean shells with the alkali sodium hydroxide. It can be used in agriculture as a seed treatment and to fight off fungal infections and is often used in the wine making industry as a chemical to prevent spoilage. In medicine it can be used in bandages to reduce bleeding and as an antibacterial agent. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 9012-76-4
Molecular Formula C56H103N9O39
Molecular Weight (g/mol) 1526.46
InChI Key FLASNYPZGWUPSU-SICDJOISSA-N
IUPAC Name methyl N-[(2S,3R,4R,5S,6R)-5-{[(2S,3R,4R,5S,6R)-3-amino-5-{[(2S,3R,4R,5S,6R)-3-amino-5-{[(2S,3R,4R,5S,6R)-3-amino-5-{[(2S,3R,4R,5S,6R)-3-amino-5-{[(2S,3R,4R,5S,6R)-3-amino-5-{[(2S,3R,4R,5S,6R)-3-amino-4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-4-hydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-4-hydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-4-hydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-4-hydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-4-hydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-2-{[(2R,3S,4R,5R,6S)-5-amino-6-{[(2R,3S,4R,5R,6R)-5-amino-4,6-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}-4-hydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}-4-hydroxy-6-(hydroxymethyl)oxan-3-yl]carbamate
SMILES COC(=O)N[C@@H]1[C@@H](O)[C@H](O[C@@H]2O[C@H](CO)[C@@H](O[C@@H]3O[C@H](CO)[C@@H](O[C@@H]4O[C@H](CO)[C@@H](O[C@@H]5O[C@H](CO)[C@@H](O[C@@H]6O[C@H](CO)[C@@H](O[C@@H]7O[C@H](CO)[C@@H](O)[C@H](O)[C@H]7N)[C@H](O)[C@H]6N)[C@H](O)[C@H]5N)[C@H](O)[C@H]4N)[C@H](O)[C@H]3N)[C@H](O)[C@H]2N)[C@@H](CO)O[C@H]1O[C@H]1[C@H](O)[C@@H](N)[C@H](O[C@H]2[C@H](O)[C@@H](N)[C@H](O)O[C@@H]2CO)O[C@@H]1CO

Specifications

CAS Max % 100%
Packaging Poly Bottle
Quantity 25 g
Grade Reagent
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.