Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Chitotriosidase Monoclonal Antibody (1D9G2,1D9G4,2B4G3)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA515482
Description
MA5-15482 targets CHIT1 in indirect ELISA, WB applications and shows reactivity with Human samples. The MA5-15482 immunogen is purified recombinant fragment of CHIT1 (aa22-137) expressed in E. Coli..
Chitotriosidase is secreted by activated human macrophages and is markedly elevated in plasma of Gaucher disease patients. The expression of chitotriosidase occurs only at a late stage of differentiation of monocytes to activated macrophages in culture. Human macrophages can synthesize a functional chitotriosidase, a highly conserved enzyme with a strongly regulated expression. This enzyme may play a role in the degradation of chitin-containing pathogens.
Specifications
Chitotriosidase | |
Monoclonal | |
Conc. Not Determined | |
Ascites with 0.03% sodium azide | |
Q13231 | |
CHIT1 | |
Purified recombinant fragment of CHIT1 (aa22-137) expressed in E. Coli. | |
RUO | |
1118 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Western Blot | |
1D9G2,1D9G4,2B4G3, 1D9G4, 2B4G3 | |
Unconjugated | |
CHIT1 | |
2300002L19Rik; CHI3; CHIT; Chit1; CHITD; chitinase 1; chitinase 1 (chitotriosidase); chitinase-1; Chitotriosidase; Chitotriosidase-1; EC 3.2.1.14; FLJ00314; MGC125322; plasma methylumbelliferyl tetra-N-acetylchitotetraoside hydrolase | |
Mouse | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction