Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Chitotriosidase Monoclonal Antibody (397504)
GREENER_CHOICE

Catalog No. PIMA524132
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524132 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524132 Supplier Invitrogen™ Supplier No. MA524132
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

In direct ELISAs, no cross-reactivity with recombinant human CHI3L1 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.

Chitotriosidase is secreted by activated human macrophages and is markedly elevated in plasma of Gaucher disease patients. The expression of chitotriosidase occurs only at a late stage of differentiation of monocytes to activated macrophages in culture. Human macrophages can synthesize a functional chitotriosidase, a highly conserved enzyme with a strongly regulated expression. This enzyme may play a role in the degradation of chitin-containing pathogens.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Chitotriosidase
Applications Immunoprecipitation, Neutralization
Classification Monoclonal
Clone 397504
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene CHIT1
Gene Accession No. Q13231
Gene Alias 2300002L19Rik; CHI3; CHIT; Chit1; CHITD; chitinase 1; chitinase 1 (chitotriosidase); chitinase-1; Chitotriosidase; Chitotriosidase-1; EC 3.2.1.14; FLJ00314; MGC125322; plasma methylumbelliferyl tetra-N-acetylchitotetraoside hydrolase
Gene Symbols CHIT1
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Chitotriosidase/CHIT1 Ala22-Asn466.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1118
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.