Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Chitotriosidase Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582395
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582395 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582395 Supplier Invitrogen™ Supplier No. PA582395
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FDDVESFKTK VSYLKQKGLG GAMVWALDLD DFAGFSCNQG RYPLIQTLRQ ELSLPYLPSG TPELEVPKPG QPSEPEHGPS PGQDTFCQGK ADGLYPNPRE RSSFYSCAAG RLFQQSCPTG LVFSNSCKCC TWN Highest antigen sequence indentity to the following orthologs: Rat - 73%, Mouse - 70%.

Chitotriosidase is secreted by activated human macrophages and is markedly elevated in plasma of Gaucher disease patients. The expression of chitotriosidase occurs only at a late stage of differentiation of monocytes to activated macrophages in culture. Human macrophages can synthesize a functional chitotriosidase, a highly conserved enzyme with a strongly regulated expression. This enzyme may play a role in the degradation of chitin-containing pathogens.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Chitotriosidase
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CHIT1
Gene Accession No. Q13231
Gene Alias 2300002L19Rik; CHI3; CHIT; Chit1; CHITD; chitinase 1; chitinase 1 (chitotriosidase); chitinase-1; Chitotriosidase; Chitotriosidase-1; EC 3.2.1.14; FLJ00314; MGC125322; plasma methylumbelliferyl tetra-N-acetylchitotetraoside hydrolase
Gene Symbols CHIT1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CHIT1. Recombinant protein control fragment (Product #RP-90229).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1118
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.