Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chk1 Antibody (2F2), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | Chk1 |
---|---|
Clone | 2F2 |
Dilution | Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:20-1:200 |
Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry |
Classification | Monoclonal |
Description
Chk1 Monoclonal antibody specifically detects Chk1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, ImmunocytochemistrySpecifications
Chk1 | |
Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:20-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
Checkpoint, S. pombe, homolog of, 1, CHK1 (checkpoint, S.pombe) homolog, CHK1 checkpoint homolog (S. pombe), CHK1serine/threonine-protein kinase Chk1, EC 2.7.11, EC 2.7.11.1 | |
A synthesized peptide derived from Human Chk1 [UniProt O14757] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
2F2 | |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, Phospho Specific, Stem Cell Markers, Tumor Suppressors | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
1111 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title