Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chk1 Antibody, Biotin, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NB100311B
Description
Chk1 Polyclonal antibody specifically detects Chk1 in Human samples. It is validated for Western BlotSpecifications
Chk1 | |
Polyclonal | |
Western Blot | |
Checkpoint, S. pombe, homolog of, 1, CHK1 (checkpoint, S.pombe) homolog, CHK1 checkpoint homolog (S. pombe), CHK1serine/threonine-protein kinase Chk1, EC 2.7.11, EC 2.7.11.1 | |
The epitope recognized by NB100-311 maps to a region between residues 250 and 300 of human Checkpoint Kinase 1 using the numbering given in entry NP_001265.1 (GeneID 1111). | |
0.1 mL | |
Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, Phospho Specific, Stem Cell Markers, Tumor Suppressors | |
1111 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
Biotin | |
PBS | |
Goat | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction