Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chk1 Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501557016
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-155-7016 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-155-7016 Supplier Bethyl Laboratories, Inc Supplier No. A300162AT
Add to Cart
Add to Cart

Goat Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

CHK1 (CHEK1) is a kinase that phosphorylates cdc25, an important phosphatase in cell cycle control, particularly for entry into mitosis. CHK1 can also phosphorylate p53 at Ser20 in vitro. Cell cycle events are regulated by the sequential activation and deactivation of cyclin dependent kinases (Cdks) and by proteolysis of cyclins. CHK1 is involved in these processes as regulators of Cdks. CHK1 function as an essential component in the G2 DNA damage checkpoint by phosphorylating Cdc25C in response to DNA damage. Phosphorylation inhibits Cdc25C activity, thereby blocking mitosis. Cdc25A, Cdc25B and Cdc25C protein tyrosine phosphatases function as mitotic activators by dephosphorylating Cdc2 p34 on regulatory tyrosine residues. CHK1 can phosphorylate Wee 1 in vitro, providing evidence that the hyperphosphorylated form of Wee 1, seen in cells delayed by CHK1 overexpression, is due to phosphorylation by CHK1. CHK1 is phosphorylated on Serine 345 (S345) in response to UV, IR and hydroxyurea (HU). CHK1 plays an essential role in the mammalian DNA damage checkpoint, embryonic development and tumor suppression. Further, CHK1 is a key mediator in the DNA damage-induced checkpoint network. CHK1 is an evolutionarily conserved protein kinase that functions to ensure genomic integrity upon genotoxic stress. When the G2 or S checkpoint is abrogated by the inhibition of CHK1, p53-deficient cancer cells undergo mitotic catastrophe and eventually apoptosis, whereas normal cells are still arrested in the G1 phase. Thus, CHK1 inhibitors can preferentially potentiate the efficacy of DNA damaging agents in cancer cells, and Chk1 is an attractive therapeutic target for cancer treatment, especially since approximately 50% of all human cancers are p53-deficient.

Specifications

Antigen Chk1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene CHEK1
Gene Accession No. O14757
Gene Alias CHEK1
Gene Symbols CHEK1
Host Species Goat
Immunogen Between 250 and 300
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1111
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.