Learn More
Chk2 Antibody (73C175.1.1), Alexa Fluor™ 532, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP227403AF532
Catalog No. NP227403A32
Only null left
Add to Cart
Description
Description
Chk2 Monoclonal specifically detects Chk2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
Chk2
Monoclonal
Alexa Fluor 532
50 mM sodium borate with 0.05% sodium azide
CHEK2
A portion of amino acids 100-150 of human Chk2 protein was used as immunogen for this antibody.
Primary
Human, Mouse
IgG1
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
73C175.1.1
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
CHK2 checkpoint homolog (S. pombe), EC 2.7.11, EC 2.7.11.1, S.pombe) homolog
Mouse
0.1 mL
11200
Store at 4°C in the dark.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.