Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chk2 Antibody (73C175.1.1) - Azide Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP227403
Description
Chk2 Monoclonal specifically detects Chk2 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Chk2 | |
Monoclonal | |
1.0 mg/ml | |
Western Blot 2-4ug/ml, Immunohistochemistry-Paraffin | |
CHK2 checkpoint homolog (S. pombe), EC 2.7.11, EC 2.7.11.1, S.pombe) homolog | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
73C175.1.1 | |
Unconjugated | |
O96017 | |
CHEK2 | |
A portion of amino acids 100-150 of human Chk2 protein was used as immunogen for this antibody. | |
0.1 mg | |
Apoptosis, Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, Phospho Specific, Stem Cell Markers | |
11200 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction