Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chk2 Antibody (73C175.1.1), Novus Biologicals™

Mouse Monoclonal Antibody
$206.50 - $426.00
Specifications
Antigen | Chk2 |
---|---|
Clone | 73C175.1.1 |
Concentration | 1.0 mg/mL |
Dilution | Western Blot 2 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10056546T
![]() |
Novus Biologicals
NB10056546SS |
0.025 mg |
Each for $206.50
|
|
|||||
NB10056546
![]() |
Novus Biologicals
NB10056546 |
0.1 mg |
Each for $426.00
|
|
|||||
Description
Chk2 Monoclonal specifically detects Chk2 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Chk2 | |
1.0 mg/mL | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Apoptosis, Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, Core ESC Like Genes, DNA Repair, Stem Cell Markers | |
O96017 | |
11200 | |
A portion of amino acids 100-150 of human Chk2 protein was used as immunogen for this antibody. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
73C175.1.1 | |
Western Blot 2 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse | |
CHK2 checkpoint homolog (S. pombe), EC 2.7.11, EC 2.7.11.1, S.pombe) homolog | |
CHEK2 | |
IgG1 | |
Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title