Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chk2 Antibody (8F12), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB100500
Description
Chk2 Monoclonal antibody specifically detects Chk2 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry, Immunofluorescence.Specifications
Chk2 | |
Monoclonal | |
1.0mg/mL | |
Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:500 | |
CHEK2 | |
Recombinant CHK2 protein | |
Protein G purified | |
RUO | |
Primary | |
Specific for human Chk2 protein. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, Immunocytochemistry, Immunofluorescence | |
8F12 | |
Unconjugated | |
CHK2 checkpoint homolog (S. pombe), EC 2.7.11, EC 2.7.11.1, S.pombe) homolog | |
Mouse | |
61 kDa | |
100 μL | |
Apoptosis, Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, Stem Cell Markers | |
11200 | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction