Learn More
Promega CHK2 Kinase Enzyme System, 10microg
Catalog No. PRV4020 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
PRV4020 10 μg
1 options
Catalog No. PRV4020 Supplier Promega Supplier No. V4020
Recombinant full-length human CHK2 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. CHK2 is rapidly phosphorylated and activated in response to replication blocks and DNA damage.
  • Easily Screen and Profile CHK2 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 10 μg
Content And Storage <-65°C