Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHKA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106290
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106290 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106290 Supplier Invitrogen™ Supplier No. PA5106290
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total Choline kinase alpha.

The major pathway for the biosynthesis of phosphatidylcholine occurs via the CDP-choline pathway. Choline kinase alpha is the initial enzyme in the sequence and may play a regulatory role. This protein also catalyzes the phosphorylation of ethanolamine.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHKA
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CHKA
Gene Accession No. O54804, P35790, Q01134
Gene Alias CHETK-alpha; CHK; CHKA; ChoK; Choline kinase alpha; choline kinase R; choline/ethanolamine kinase alpha; CK; CK/EK-alpha; CKI; Ckr; CK-R; EK; ethanolamine kinase; EtnK-alpha
Gene Symbols CHKA
Host Species Rabbit
Immunogen A synthesized peptide derived from human CHKA(Accession P35790), corresponding to amino acid residues L313-R363.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1119, 12660, 29194
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.