Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CHL-1/L1CAM-2 Antibody, Janelia Fluor™ 646, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298946JF646
Description
CHL-1/L1CAM-2 Polyclonal antibody specifically detects CHL-1/L1CAM-2 in Human samples. It is validated for ELISASpecifications
CHL-1/L1CAM-2 | |
Polyclonal | |
ELISA | |
CALLClose homolog of L1, cell adhesion molecule with homology to L1CAM (close homolog of L1), cell adhesion molecule with homology to L1CAM (close homologue of L1), FLJ44930, L1CAM2L1 cell adhesion molecule 2, MGC132578, neural cell adhesion molecule L1-like protein | |
Produced in rabbits immunized with purified, recombinant Human CHL-1/L1CAM-2 (Accession#: AAI04919.1; Met 1-Gln 1080) | |
0.1 mL | |
Primary | |
Human | |
Purified |
ELISA | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
10752 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction