Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CHL-1/L1CAM-2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | CHL-1/L1CAM-2 |
---|---|
Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
CHL-1/L1CAM-2 Polyclonal specifically detects CHL-1/L1CAM-2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
CHL-1/L1CAM-2 | |
Polyclonal | |
Rabbit | |
Human | |
CALLClose homolog of L1, cell adhesion molecule with homology to L1CAM (close homolog of L1), cell adhesion molecule with homology to L1CAM (close homologue of L1), FLJ44930, L1CAM2L1 cell adhesion molecule 2, MGC132578, neural cell adhesion molecule L1-like protein | |
CHL1 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
10752 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:CEFFASPEAVVSWQKVEEVKPLEGRRYHIYENGTLQINRTTEEDAGSYSCWVENAIGKTAVTANLDIRNATKLRVSPKNPRIPKLHMLELHCESKCDSHLKHSLKLSWSKDGEAFEINGTEDGRIIIDGANLTISNVTLEDQGIYCCS | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title