Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115014
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115014 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115014 Supplier Invitrogen™ Supplier No. PA5115014
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total CHL1.

The protein encoded by this gene is a member of the L1 gene family of neural cell adhesion molecules. It is a neural recognition molecule that may be involved in signal transduction pathways. The deletion of one copy of this gene may be responsible for mental defects in patients with 3p- syndrome. Several alternatively spliced transcript variants of this gene have been described, but their full length nature is not known.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHL1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene CHL1
Gene Accession No. O00533, P70232
Gene Alias A530023M13Rik; AI465420; CALL; cell adhesion molecule L1 like; cell adhesion molecule L1-like; cell adhesion molecule with homology to L1CAM; cell adhesion molecule with homology to L1CAM (close homolog of L1); cell adhesion molecule with homology to L1CAM (close homologue of L1); Chl1; Chl1-like protein; close homolog of L1; L1 cell adhesion molecule 2; L1CAM2; LICAM2; neural cell adhesion molecule L1-like protein; Processed neural cell adhesion molecule L1-like protein
Gene Symbols CHL1
Host Species Rabbit
Immunogen A synthesized peptide derived from human CHL1(Accession O00533), corresponding to amino acid residues G1168-A1208.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10752, 12661, 89828
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.